Anti-NDUFB7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6658A
89391-802EA 589.74 USD
89391-802
Anti-NDUFB7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFB7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR