Anti-FGF18 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6643A
89391-762EA 623.77 USD
89391-762
Anti-FGF18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR