Anti-dj-1beta Rabbit Polyclonal Antibody

Supplier: Abgent
AP6417A
89391-368EA 589.74 USD
89391-368
Anti-dj-1beta Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: dj-1beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophohila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR