Anti-GPR81 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6362D
89391-284EA 589.74 USD
89391-284
Anti-GPR81 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Gpr81
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR