Anti-IL1RL2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6355B
89391-252EA 619.38 USD
89391-252
Anti-IL1RL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1RL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR