Anti-RRAGA Rabbit Polyclonal Antibody

Supplier: Abgent
AP5772B
89390-264EA 589.74 USD
89390-264
Anti-RRAGA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RRAGA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR