Anti-CCNYL1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5730A
89390-180EA 619.21 USD
89390-180
Anti-CCNYL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCNYL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR