Anti-C9orf24 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5603C
89389-928EA 589.74 USD
89389-928
Anti-C9orf24 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C9orf24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR