Anti-GNE Rabbit Polyclonal Antibody

Supplier: Abgent
AP5318B
89389-368EA 619.21 USD
89389-368
Anti-GNE Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR