Anti-ZBTB5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5302A
89389-336EA 619.21 USD
89389-336
Anti-ZBTB5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZBTB5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR