Anti-CMPK2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5206C
89389-144EA 619.21 USD
89389-144
Anti-CMPK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CMPK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR