Anti-CK068 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5204B
89389-140EA 589.74 USD
89389-140
Anti-CK068 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CK068
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR