Anti-CA052 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5196B
89389-124EA 589.56 USD
89389-124
Anti-CA052 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CA052
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR