Anti-ATP6V1H Rabbit Polyclonal Antibody

Supplier: Abgent
AP5182B
89389-096EA 619.21 USD
89389-096
Anti-ATP6V1H Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATP6V1H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR