Anti-CHRNA3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP5022A
89388-770EA 619.55 USD
89388-770
Anti-CHRNA3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHRNA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR