Anti-FUT6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP4925C
89388-574EA 619.21 USD
89388-574
Anti-FUT6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FUT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR