Anti-CENPN Rabbit Polyclonal Antibody

Supplier: Abgent
AP4913C
89388-550EA 589.74 USD
89388-550
Anti-CENPN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPN
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR