Anti-PDHE1A Rabbit Polyclonal Antibody

Supplier: Abgent
AP3758A
89388-130EA 632.53 USD
89388-130
Anti-PDHE1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDHE1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR