Anti-MAP2K2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3489A
89387-610EA 590.05 USD
89387-610
Anti-MAP2K2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR