Anti-MAP4K4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3444A
89387-522EA 619.55 USD
89387-522
Anti-MAP4K4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP4K4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR