Anti-HIST1H3B Rabbit Polyclonal Antibody

Supplier: Abgent
AP3115A
89387-052EA 615.7 USD
89387-052
Anti-HIST1H3B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIST1H3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR