Anti-P4HB Rabbit Polyclonal Antibody

Supplier: Abgent
AP2911B
89386-780EA 619.55 USD
89386-780
Anti-P4HB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: P4HB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR