Anti-HSPA2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2858A
89386-608EA 589.74 USD
89386-608
Anti-HSPA2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSPA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR