Anti-DHCR24 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2840C
89386-552EA 619.21 USD
89386-552
Anti-DHCR24 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DHCR24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR