Anti-GADD45A Rabbit Polyclonal Antibody

Supplier: Abgent
AP2785A
89386-374EA 619.21 USD
89386-374
Anti-GADD45A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GADD45A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR