Anti-LLGL1, LLGL2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2198A
89385-802EA 615.89 USD
89385-802
Anti-LLGL1, LLGL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LLGL1, LLGL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR