Anti-dcr-1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1961C
89385-220EA 619.21 USD
89385-220
Anti-dcr-1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: dcr-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: C. Elegans
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR