Anti-SFRS12 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1944A
89385-160EA 589.74 USD
89385-160
Anti-SFRS12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SFRS12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR