Anti-CSE1L Rabbit Polyclonal Antibody

Supplier: Abgent
AP1935A
89385-132EA 615.89 USD
89385-132
Anti-CSE1L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSE1L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR