Anti-FUBP3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1916A
89385-070EA 589.74 USD
89385-070
Anti-FUBP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FUBP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR