Anti-ATG3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1807E
89384-834EA 589.74 USD
89384-834
Anti-ATG3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR