Anti-CDH7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1471A
89384-450EA 619.21 USD
89384-450
Anti-CDH7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDH7
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR