Anti-DPF2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1451A
89384-372EA 590.05 USD
89384-372
Anti-DPF2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DPF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR