Anti-Nek8 Rabbit Polyclonal Antibody

Supplier: Abgent
AP13927B
89383-718EA 619.21 USD
89383-718
Anti-Nek8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Nek8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse (Predicted)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR