Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP13835A
89383-534EA 619.21 USD
89383-534
Anti-CASP6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR