Anti-CD3E Rabbit Polyclonal Antibody

Supplier: Abgent
AP13531B
89382-922EA 619.21 USD
89382-922
Anti-CD3E Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR