Anti-WDR12 Rabbit Polyclonal Antibody

Supplier: Abgent
AP13076C
89382-606EA 619.21 USD
89382-606
Anti-WDR12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WDR12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR