Anti-C9orf72 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12928B
89382-292EA 615.76 USD
89382-292
Anti-C9orf72 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C9orf72
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR