Anti-GCAT Rabbit Polyclonal Antibody

Supplier: Abgent
AP12897A
89382-232EA 619.21 USD
89382-232
Anti-GCAT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GCAT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR