Anti-GPS1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12706C
89381-834EA 619.21 USD
89381-834
Anti-GPS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human (Predicted), Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR