Anti-CDKN2B Rabbit Polyclonal Antibody

Supplier: Abgent
AP12629A
89381-678EA 619.21 USD
89381-678
Anti-CDKN2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDKN2B
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR