Anti-RPRML Rabbit Polyclonal Antibody

Supplier: Abgent
AP12602B
89381-622EA 589.74 USD
89381-622
Anti-RPRML Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPRML
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR