Anti-GOLGA8F Rabbit Polyclonal Antibody

Supplier: Abgent
AP12408A
89381-200EA 589.74 USD
89381-200
Anti-GOLGA8F Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GOLGA8F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR