Anti-HAX1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12311B
89380-990EA 619.21 USD
89380-990
Anti-HAX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR