Anti-CEACAM18 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12282A
89380-926EA 626.74 USD
89380-926
Anti-CEACAM18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CEACAM18
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR