Anti-CSAD Rabbit Polyclonal Antibody

Supplier: Abgent
AP11783C
89379-888EA 619.21 USD
89379-888
Anti-CSAD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSAD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR