Anti-TMEM150B Rabbit Polyclonal Antibody

Supplier: Abgent
AP11399A
89379-110EA 619.21 USD
89379-110
Anti-TMEM150B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TMEM150B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR