Anti-ZDHHC19 Rabbit Polyclonal Antibody

Supplier: Abgent
AP11353B
89379-018EA 589.74 USD
89379-018
Anti-ZDHHC19 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZDHHC19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR