Anti-CYB5D2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP11281B
89378-874EA 619.21 USD
89378-874
Anti-CYB5D2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYB5D2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR