Anti-CYP2B6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP11104C
89378-496EA 619.38 USD
89378-496
Anti-CYP2B6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP2B6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR